LIF432341543 Life Chemicals Inc.

4-(aminomethyl)-2-(2-(pyrrolidin-1-yl)ethyl)phthalazin-1(2H)-one dihydrochloride

  • MDL number MFCD28016969
  • Cas Number : 1955530-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart