LIF432341551 Life Chemicals Inc.

2-chloro-6-(methylsulfonyl)benzo[d]thiazole

  • MDL number MFCD13190213
  • Cas Number : 3622-29-5
  • Molecular Weight : 247.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart