LIF432341552 Life Chemicals Inc.

2-chloro-4-(methylthio)benzo[d]thiazole

  • MDL number MFCD26940469
  • Cas Number : 1706440-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart