LIF432341555 Life Chemicals Inc.

3-(5-azetidin-3-yl-1,2,4-oxadiazol-3-yl)pyridine dihydrochloride

  • MDL number MFCD16652926
  • Cas Number : 1426290-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart