LIF432341558 Life Chemicals Inc.

N-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD28035754
  • Cas Number : 1820717-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart