LIF432341559 Life Chemicals Inc.

N-cyclopentylpiperidine-4-carboxamide hydrochloride

  • MDL number MFCD13562552
  • Cas Number : 1220038-00-5
  • Molecular Weight : 232.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart