LIF432341562 Life Chemicals Inc.

5-azetidin-3-yl-3-benzyl-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD16652977
  • Cas Number : 1351620-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart