LIF432341563 Life Chemicals Inc.

5-azetidin-3-yl-3-(2-methoxyphenyl)-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD16652983
  • Cas Number : 1426291-17-9
  • Molecular Weight : 267.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart