LIF432341565 Life Chemicals Inc.

4-[(benzyloxy)methyl]piperidine hydrochloride

  • MDL number MFCD09879195
  • Cas Number : 301226-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart