LIF432341569 Life Chemicals Inc.

5-(4-(ethylthio)benzyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD20229783
  • Cas Number : 1251682-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart