LIF432341570 Life Chemicals Inc.

5-(4-(methylsulfonyl)benzyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD22587576
  • Cas Number : 1257553-09-5
  • Molecular Weight : 253.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart