LIF432341576 Life Chemicals Inc.

1-((3,5-dimethyl-1H-pyrazol-4-yl)sulfonyl)piperazine hydrochloride

  • MDL number MFCD10042187
  • Cas Number : 1240238-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart