LIF432341577 Life Chemicals Inc.

1-benzyl-2-piperazin-1-yl-1H-benzimidazole dihydrochloride

  • MDL number MFCD22587621
  • Cas Number : 1351654-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart