LIF432341579 Life Chemicals Inc.

2-aminobenzo[d]thiazole-6-sulfonic acid

  • MDL number MFCD00071759
  • Cas Number : 21951-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart