LIF432341581 Life Chemicals Inc.

methyl 2-(2-imino-6-methylbenzo[d]thiazol-3(2H)-yl)acetate hydrobromide

  • MDL number MFCD26940477
  • Cas Number : 1820588-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart