LIF432341582 Life Chemicals Inc.

3-(prop-2-yn-1-yl)benzo[d]thiazol-2(3H)-imine hydrobromide

  • MDL number MFCD26940480
  • Cas Number : 3708-59-6
  • Molecular Weight : 269.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart