LIF432341583 Life Chemicals Inc.

6-ethyl-3-(prop-2-yn-1-yl)benzo[d]thiazol-2(3H)-imine hydrobromide

  • MDL number MFCD26940482
  • Cas Number : 1820586-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart