LIF432341584 Life Chemicals Inc.

7-(prop-2-yn-1-yl)-[1,3]dioxolo[4',5':4,5]benzo[1,2-d]thiazol-6(7H)-imine hydrobromide

  • MDL number MFCD26940484
  • Cas Number : 1820590-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart