LIF432341594 Life Chemicals Inc.

methyl 2-(6-butyl-2-iminobenzo[d]thiazol-3(2H)-yl)acetate hydrobromide

  • MDL number MFCD22587732
  • Cas Number : 1351596-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart