LIF432341595 Life Chemicals Inc.

2-amino-N,6-dimethyl-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide

  • MDL number MFCD00782330
  • Cas Number : 915920-53-5
  • Molecular Weight : 224.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart