LIF432341598 Life Chemicals Inc.

5-azetidin-3-yl-3-(3-thienyl)-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD24387581
  • Cas Number : 1426290-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart