LIF432341600 Life Chemicals Inc.

5-(azetidin-3-yl)-3-(furan-2-yl)-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD28016970
  • Cas Number : 1820674-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart