LIF432341604 Life Chemicals Inc.

1-(4-(trifluoromethyl)benzo[d]thiazol-2-yl)azetidin-3-ol

  • MDL number MFCD22587766
  • Cas Number : 1421468-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart