LIF432341611 Life Chemicals Inc.

2-amino-N-methyl-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide

  • MDL number MFCD00782672
  • Cas Number : 666715-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart