LIF432341614 Life Chemicals Inc.

3-methyl-4-(1-methyl-1,2,3,6-tetrahydropyridin-4-yl)isoxazol-5-amine

  • MDL number MFCD20700588
  • Cas Number : 88786-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart