LIF432341620 Life Chemicals Inc.

ethyl 1-isopropyl-5-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD08700679
  • Cas Number : 50920-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart