LIF432341629 Life Chemicals Inc.

ethyl 2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate

  • MDL number MFCD18453645
  • Cas Number : 74004-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart