LIF432341645 Life Chemicals Inc.

2-(4-ethoxyphenyl)acetyl chloride

  • MDL number MFCD00995155
  • Cas Number : 10368-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart