LIF432341646 Life Chemicals Inc.

2-(2,5-dimethylphenyl)acetyl chloride

  • MDL number MFCD12024950
  • Cas Number : 55312-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart