LIF432341651 Life Chemicals Inc.

1-((4-fluorophenyl)sulfonyl)piperidine-3-carbonyl chloride

  • MDL number MFCD26940509
  • Cas Number : 1706435-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart