LIF432341652 Life Chemicals Inc.

5-(benzo[d]thiazol-2-yl)thiophene-2-carbonyl chloride

  • MDL number MFCD26940510
  • Cas Number : 1706462-04-5
  • Molecular Weight : 279.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart