LIF432341653 Life Chemicals Inc.

1-methyl-1H-pyrazole-3-carbonyl chloride

  • MDL number MFCD03419367
  • Cas Number : 84547-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart