LIF432341664 Life Chemicals Inc.

4-amino-1-(3,4-dimethylphenyl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD11573341
  • Cas Number : 1177279-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart