LIF432341665 Life Chemicals Inc.

4-Amino-1-(2,6-difluorobenzyl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD09743241
  • Cas Number : 1172854-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart