LIF432341667 Life Chemicals Inc.

3,4-dihydrospiro[chromene-2,4'-piperidine] hydrochloride

  • MDL number MFCD16661064
  • Cas Number : 400729-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart