LIF432341668 Life Chemicals Inc.

1-{[1-(4-methoxyphenyl)-1H-tetrazol-5-yl]methyl}piperazine dihydrochloride

  • MDL number MFCD16661031
  • Cas Number : 1351643-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart