LIF432341677 Life Chemicals Inc.

methyl amino(3-chlorophenyl)acetate hydrochloride

  • MDL number MFCD16661102
  • Cas Number : 1351586-91-8
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart