LIF432341681 Life Chemicals Inc.

3-(tert-butoxycarbonyl)-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid

  • MDL number MFCD03644281
  • Cas Number : 301155-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart