LIF432341686 Life Chemicals Inc.

{[4-(4-methoxyphenyl)-4H-1,2,4-triazol-3-yl]methyl}amine dihydrochloride

  • MDL number MFCD22587831
  • Cas Number : 1351609-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart