LIF432341688 Life Chemicals Inc.

{[4-phenyl-5-(trifluoromethyl)-4H-1,2,4-triazol-3-yl]methyl}amine hydrochloride

  • MDL number MFCD22587833
  • Cas Number : 1351659-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart