LIF432341689 Life Chemicals Inc.

5-(aminomethyl)thiophene-2-carbonitrile hydrochloride

  • MDL number MFCD06658320
  • Cas Number : 172349-10-9
  • Molecular Weight : 174.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart