LIF432341690 Life Chemicals Inc.

4-phenyl-2-pyrrolidin-2-yl-1H-imidazole dihydrochloride

  • MDL number MFCD17480426
  • Cas Number : 1258640-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart