LIF432341694 Life Chemicals Inc.

Potassium 5-methyl-1,3,4-oxadiazole-2-carboxylate

  • MDL number MFCD08060085
  • Cas Number : 888504-28-7
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart