LIF432341697 Life Chemicals Inc.

(4-pyrrolidin-1-ylbut-2-yn-1-yl)amine dihydrochloride

  • MDL number MFCD22587855
  • Cas Number : 147555-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart