LIF432341698 Life Chemicals Inc.

4-(azepan-1-yl)but-2-yn-1-amine dihydrochloride

  • MDL number MFCD22587856
  • Cas Number : 1396770-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart