LIF432341699 Life Chemicals Inc.

N1-benzyl-N1-methylbut-2-yne-1,4-diamine dihydrochloride

  • MDL number MFCD22587859
  • Cas Number : 1396761-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart