LIF432341700 Life Chemicals Inc.

1-(1-methylpiperidin-4-yl)-5-oxopyrrolidine-3-carboxylic acid hydrate

  • MDL number MFCD22587879
  • Cas Number : 1396886-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart