LIF432341702 Life Chemicals Inc.

3-methyl-5-oxo-5,8-dihydro[1,2,4]triazolo[4,3-a]pyrimidine-6-carboxylic acid hydrate

  • MDL number MFCD22587880
  • Cas Number : 1396784-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart