LIF432341713 Life Chemicals Inc.

(2-methoxypyridin-3-yl)methanamine

  • MDL number MFCD09740382
  • Cas Number : 354824-19-4
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart