LIF432341716 Life Chemicals Inc.

3-(aminomethyl)-N,N-dimethylpyridin-2-amine

  • MDL number MFCD09731450
  • Cas Number : 354824-09-2
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart